EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Evaluation of Ricide-P pesticide toxicity using the experimental model mother-fetus-newborn].
PMID 8244054
Abstract
Necessity of the mother-fetus-newborn model usage in toxicological experiments for the xenobiotics environmental content standardization was shown in the toxicological study of Ricid-P pesticide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45814
Iprobenfos, PESTANAL®, analytical standard
C13H21O3PS