EMAIL THIS PAGE TO A FRIEND
Biochemical medicine and metabolic biology
A profile of cerebral and hepatic carnitine, ammonia, and energy metabolism in a model of organic aciduria: BALB/cByJ mouse with short-chain acyl-CoA dehydrogenase deficiency.
PMID 8260192
Abstract
Spontaneous animal models of inborn errors of metabolism are valuable tools for defining the pathogenesis of these disorders and also the mechanism of various therapeutic approaches. In the present study, we have employed BALB/cByJ mice with an autosomal recessive deficiency of short-chain acyl-CoA dehydrogenase (SCAD). These animals were characterized by a marked urinary excretion of ethylmalonic and methylsuccinic acids along with butyrylglycine. Using adult homozygous mice we have studied the basic cerebral and hepatic profile of carnitine, ammonia, and energy metabolism. The effects of fasting and a short-term supplement of L-carnitine have been evaluated in comparison with control BALB/cJ mice. The mutant mice had low levels of acetyl-CoA and high levels of lactate compared to control mice. Fasting aggravated this condition by further decreasing acetyl-CoA and increasing lactate levels in the mutant mice. Free carnitine levels were significantly decreased in liver with fasting. Long-chain acylcarnitines were significantly lower in the brain of mutant mice. A short-term supplementation of L-carnitine resulted in general increases of carnitine levels in liver and muscle, but they still remained lower in mutant BALB/cByJ mice as compared to control BALB/cJ mice. L-Carnitine treatment increased cerebral CoA-SH levels and both hepatic and cerebral acetyl-CoA levels in mutant mice. Hyperammonemia which has been described frequently in acyl-CoA dehydrogenase deficiencies was not observed in adult BALB/cByJ mice. This could be due to a rapid conjugation of butyryl-CoA with glycine by an increased activity of glycine N-acyltransferase.