EMAIL THIS PAGE TO A FRIEND
Ukrainskii biokhimicheskii zhurnal (1978)
[Antioxidant activity of a new derivative of 4-hydrazinoquinazoline in experimental cerebral hypoxia].
PMID 8291133
Abstract
It is established that KS-79 compound possesses the high level of antioxidant activity and realizes its antioxidant effect at initial stages of free-radical hydrogen peroxide oxidation of lipids. Due to this ability it differs from the oxidants of direct-type action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBR00329
4-Hydrazinoquinazoline, AldrichCPR
C8H8N4