EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Continuous electrophoretic separations in narrow channels coupled to small-bore capillaries.
PMID 8291681
Abstract
Continuous zone electrophoretic separations in channels have been demonstrated. This new technique has the potential to continuously sample and separate analytes from volume-limited microenvironments. A small-bore capillary is used to electrophoretically sample, but not separate, a mixture of dansylated amino acids. The capillary is coupled to a quartz channel structure in a manner which allows continuous injection of the sampled material into the channel. The channel functions to continuously separate the sampled material via electrophoresis. A laser-induced fluorescence detection scheme, which involves two fiber optic arrays situated at the channel exit, monitors eluting analytes. A continuous separation of dansylated amino acids on the time scale of a few minutes demonstrates the utility of the technique. Sampling has been performed continuously up to 400 s, and initial detection limits are approximately 30 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9006
Nε-Dansyl-L-lysine
C18H25N3O4S