EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Synthesis of 3-episiastatin B analogues having anti-influenza virus activity.
PMID 8294247
Abstract
Two epimers of siastatin B, 3-episiastatin B (3) and 3,4-diepisiastatin B (4), were obtained by the chemical modification of siastatin B. Compound 3 showed marked inhibitory activity against influenza virus neuraminidases and significant inhibition of influenza virus infection in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8063
Siastatin B, lyophilized powder, from microbial
C8H14N2O5