EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Glyceryl ether monooxygenase [EC 1.14.16.5]: stoichiometry and inhibition.
PMID 8304231
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B402
Batyl alcohol, 99%
C21H44O3
D6277
N-Decanoyl-N-methylglucamine, ≥98% (GC)
C17H35NO6