EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Antimalarial activity in vitro of the glyoxalase I inhibitor diester, S-p-bromobenzylglutathione diethyl ester.
PMID 8304987
Abstract
S-p-Bromobenzylglutathione diethyl ester induced toxicity in the malarial parasite Plasmodium falciparum in infected human red blood cells in culture. The median inhibitory concentration, IC50, was 4.77 +/- 0.12 microM (N = 10) for incorporation of [3H]hypoxanthine in nucleotide synthesis and 5.20 +/- 0.1 microM (N = 10) for incorporation of [14C]isoleucine into protein. The prospective mechanism of action is inhibition of glyoxalase I by the de-esterified metabolite, S-p-bromobenzylglutathione, and accumulation of the cytotoxic substrate methylglyoxal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08357 Glutathione reduced diethyl ester, BioChemika
C14H25N3O6S