EMAIL THIS PAGE TO A FRIEND
Biophysical journal
Covalent binding of biological samples to solid supports for scanning probe microscopy in buffer solution.
PMID 8312482
Abstract
Scanning force microscopy allows imaging of biological molecules in their native state in buffer solution. To this end samples have to be fixed to a flat solid support so that they cannot be displaced by the scanning tip. Here we describe a method to achieve the covalent binding of biological samples to glass surfaces. Coverslips were chemically modified with the photoactivatable cross-linker N-5-azido-2-nitrobenzoyloxysuccinimide. Samples are squeezed between derivatized coverslips and then cross-linked to the glass surface by irradiation with ultraviolet light. Such samples can be imaged repeatedly by the scanning force microscope without loss of image quality, whereas identical but not immobilized samples are pushed away by the stylus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3282
5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide ester, ≥95%, powder
C11H7N5O6