EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Quantitative determination of chitosan and the percentage of free amino groups.
PMID 8317699
Abstract
Chitosans were quantified with ninhydrin, a reagent normally used for recognizing and quantifying amino groups. The reaction was time dependent and there was no effect when different acids were added to the mixture. This method was used to determine the percentage of free amino groups in chitosans of different origins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151173
Ninhydrin, ACS reagent
C9H6O4