EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The isolation and identification of 6-hydroxycyclohepta-1,4-dione as a novel intermediate in the bacterial degradation of atropine.
PMID 8328951
Abstract
Growth of Pseudomonas AT3 on the alkaloid atropine as its sole source of carbon and nitrogen is nitrogen-limited and proceeds by degradation of the tropic acid part of the molecule, with the metabolism of the tropine being limited to the point of release of its nitrogen. A nitrogen-free compound accumulated in the growth medium and was isolated and identified as 6-hydroxycyclohepta-1,4-dione. This novel compound is proposed as an intermediate in tropine metabolism. It served as a growth substrate for the organism and was also the substrate for an NAD(+)-linked dehydrogenase present in cell extracts. The enzyme was induced during the tropine phase of diauxic growth on atropine or during growth on tropine alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T89206
Tropic acid, 98%
C9H10O3
93550
Tropine, ≥97.0% (NT)
C8H15NO