EMAIL THIS PAGE TO A FRIEND
The Journal of the Association of Physicians of India
Gastric mucosa in acute viral hepatitis.
PMID 8335612
Abstract
In acute viral hepatitis, gastric biopsy and augmented histamine test were performed in 63 patients. Gastric histology was abnormal in 12 of 63 (19%) patients; 3 with superficial and 9 with atrophic gastritis. Maximal acid output was less than 10 mEq/h in 35 of 63 (55.5%) patients. Acid load tests were performed in 23 patients measuring H+, K+, Na+ fluxes; the results showed increased loss of acid from lumen to mucosa in 16 of 23 (69.6%) patients, indicating damage to the gastric mucosal barrier.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53310
Histamine bisphosphate monohydrate, ≥99.0% (NT)
C5H9N3 · 2H3PO4 · H2O
H7375
Histamine bisphosphate monohydrate
C5H9N3 · 2H3PO4 · H2O