EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
The pharmacokinetics of sulfachlorpyridazine in channel catfish, Ictalurus punctatus.
PMID 8345573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9882
Sulfachloropyridazine
C10H9ClN4O2S
46778
Sulfachloropyridazine, VETRANAL®, analytical standard
C10H9ClN4O2S