EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Gene dose of apolipoprotein E type 4 allele and the risk of Alzheimer's disease in late onset families.
PMID 8346443
Abstract
The apolipoprotein E type 4 allele (APOE-epsilon 4) is genetically associated with the common late onset familial and sporadic forms of Alzheimer's disease (AD). Risk for AD increased from 20% to 90% and mean age at onset decreased from 84 to 68 years with increasing number of APOE-epsilon 4 alleles in 42 families with late onset AD. Thus APOE-epsilon 4 gene dose is a major risk factor for late onset AD and, in these families, homozygosity for APOE-epsilon 4 was virtually sufficient to cause AD by age 80.