EMAIL THIS PAGE TO A FRIEND
Medicine and science in sports and exercise
Effect of training and anabolic-androgenic steroids on drug metabolism in rat liver.
PMID 8350704
Abstract
The effects of anabolic-androgenic steroid administration and exercise training on various aspects of hepatic function were investigated in sedentary and trained (treadmill for 12 wk) male and female rats treated orally with fluoxymesterone or methylandrostanolone (2 mg.kg-1 body weight, 5 d.wk-1 for 8 wk). The mean values of serum alanine aminotransferase, aspartate aminotransferase, alkaline phosphatase, total and direct bilirubin, and total- and high-density lipoprotein-cholesterol remained within normal range in all groups of male animals. The same is true for female rats, except for an increase in alkaline phosphatase activity in the steroid-treated groups. Hepatic microsomal aniline p-hydroxylase activity was reduced in male and increased in female rats by either steroid, whereas no significant effect was detected on 7-ethoxycoumarin deethylase activity. The levels of cytochrome P-450 and cytochrome b5 were markedly decreased by the anabolic-androgenic steroid treatment in male rat microsomes, but neither the steroid administration nor exercise training induced significant changes in the cytochrome levels of female rat livers. Taking into account the significant increase in microsomal protein yield elicited by fluoxymesterone or methylandrostanolone treatment both in males and females, it is noteworthy that the total monooxygenase activities and cytochrome P-450 content, expressed on a per gram liver basis, were significantly increased in female whereas they were apparently unchanged in male rats. In conclusion, the present data show that the prolonged ingestion of high doses of anabolic-androgenic steroids, either with or without concurrent exercise training, can modify in a sex-dependent manner the capacity of rat liver to metabolize drugs without affecting classical serum indicators of hepatic function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5626
17α-Methylandrostan-17β-ol-3-one, analytical standard
C20H32O2