EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Binding of methylglyoxal to albumin and formation of fluorescent adducts. Inhibition by arginine, N-alpha-acetylarginine and aminoguanidine.
PMID 8359423
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3133
Nα-Acetyl-L-arginine
C8H16N4O3