EMAIL THIS PAGE TO A FRIEND
Avian diseases
Coumafuryl (Fumarin) toxicity in chicks.
PMID 8363526
Abstract
Coumafuryl (Fumarin) toxicity was diagnosed in chickens less than 1 week of age. Mortality rate was 100%. Necropsy showed crops and gizzards to be full of feed. There was diffuse hemorrhage and unclotted blood in the abdomen and thorax. Histological examination showed congestion and hemorrhage. Chemical analysis of crop and gizzard contents contained approximately 340 ppm coumafuryl. The source of the coumafuryl was found to be the wood-straw mats in the chick boxes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34324
Coumafuryl, PESTANAL®, analytical standard
C17H14O5