EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact dermatitis from thioxolone.
PMID 8365186
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217077
6-Hydroxy-1,3-benzoxathiol-2-one, ≥98%
C7H4O3S