EMAIL THIS PAGE TO A FRIEND
The New Zealand medical journal
Dyestuffs and bladder cancer.
PMID 8367102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1144
Chlorazol Black
C34H25N9Na2O7S2