EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Evaluation of indoxyl-beta-D-glucuronide as a chromogen in media specific for Escherichia coli.
PMID 8368860
Abstract
Indoxyl-beta-D-glucuronide (indoxyl) was evaluated as a specific chromogen for detection of Escherichia coli by the membrane filter method. In all, 413 colonies were tested from the indoxyl-supplemented media, yielding 93.3% confirmation, as E. coli. Compared with the indoxyl medium, other media gave either much lower recovery with high verification or equal recovery with poor verification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7638
Indoxyl β-D-glucuronide cyclohexylammonium salt
C14H15NO7 · C6H13N