EMAIL THIS PAGE TO A FRIEND
Biochemistry
Reductive dehalogenation by cytochrome P450CAM: substrate binding and catalysis.
PMID 8369306
Abstract
Biological reductive dehalogenation reactions are important in environmental detoxification of organohalides. Only scarce information is available on the enzymology underlying these reactions. Cytochrome P450CAM with a known X-ray structure and well-studied oxygenase reaction cycle, has been studied for its ability to reduce carbon-halogen bonds under anaerobic conditions. The reductive reactions functioned with NADH and the physiological electron-transfer proteins or by using artificial electron donors to reduce cytochrome P450CAM. Halogenated methane and ethane substrates were transformed by a two-electron reduction and subsequent protonation, beta-elimination, or alpha-elimination to yield alkanes, alkene, or carbene-derived products, respectively. Halogenated substrates bound to the camphor binding site as indicated by saturable changes in the Fe(III)-heme spin state upon substrate addition. Hexachloromethane was bound with a dissociation constant (KD) of 0.7 microM and caused > 95% shift from low- to high-spin iron. Ethanes bearing fewer chlorine substituents were bound with increasing dissociation constants and gave lesser degrees of iron spin-state change. Camphor competitively inhibited hexachloroethane reduction with an inhibitor constant (KI) similar to the dissociation constant for camphor (KI = KD = 0.9 microM). Rate determinations with pentachloroethane indicated a 100-fold higher enzyme V/K compared to the second-order rate constant for hematin free in solution. These studies on substrate binding and catalysis will help reveal how biological systems enzymatically reduce carbon-halogen bonds in the environment.