EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Fluorinated probes to measure carboxylesterase activities using 19F NMR spectroscopy: application to erythrocytes and Helicobacter pylori.
PMID 8373162
Abstract
Eight fluorinated compounds were tested as putative probes to measure carboxylesterase activity employing 19F nuclear magnetic resonance spectroscopy. The method takes advantage of the sensitivity of fluorine resonances to the changes in the chemical bonding in the covalent structures where they are located. Determination of the kinetic parameters of ethyl fluoroacetate and diethyl fluoromalonate in hemolysates showed that these probes were well suited to study carboxylesterase activities in complex systems the potential of these probes for noninvasive applications was demonstrated in measurements of carboxylesterase kinetic parameters in intact erythrocytes. The presence of carboxylesterases was established in several strains of the bacterium Helicobacter pylori employing 19F nuclear magnetic resonance spectroscopy, and the kinetic parameters of these enzymes for ethyl fluoroacetate and diethyl fluoromalonate were measured.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
285587
Diethyl fluoromalonate, 97%
C7H11FO4