EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Highly sensitive determination of imidapril, a new angiotensin I-converting enzyme inhibitor, and its active metabolite in human plasma and urine using high-performance liquid chromatography with fluorescent labelling reagent.
PMID 8376543
Abstract
A method for the simultaneous determination of imidapril and its active metabolite in human plasma and urine has been developed using high-performance liquid chromatography with a fluorescent labelling reagent, 9-anthryldiazomethane. Imidapril and its active metabolite were extracted from human plasma and urine using a solid-phase extraction cartridge (Bond Elut C18). Two compounds in the eluate were derivatized with 9-anthryldiazomethane and purified with a solid-phase extraction cartridge (Bond Elut SI). The derivatives were analysed using high-performance liquid chromatography with fluorometry. The detection limits of imidapril and its active metabolite were 0.2 ng/ml in plasma and 10 ng/ml in urine. This method could be applied to the pharmacokinetic study of imidapril.