EMAIL THIS PAGE TO A FRIEND
Annals of clinical biochemistry
Glycated 6-aminohexanoic acid--an improved calibrator for the serum fructosamine assay.
PMID 8379656
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6149
1-Deoxy-1-morpholino-D-fructose
C10H19NO6