EMAIL THIS PAGE TO A FRIEND
Cell
cPLA2 is phosphorylated and activated by MAP kinase.
PMID 8381049
Abstract
Treatment of cells with agents that stimulate the release of arachidonic acid causes increased serine phosphorylation and activation of cytosolic phospholipase A2 (cPLA2). Here we report that cPLA2 is a substrate for mitogen-activated protein (MAP) kinase. Moreover, phosphorylation by MAP kinase increases the enzymatic activity of cPLA2. The site of cPLA2 phosphorylation by MAP kinase, Ser-505, is identical to the major site of cPLA2 phosphorylation observed in phorbol ester-treated cells. Replacement of Ser-505 with Ala resulted in a mutant cPLA2 that is not a substrate for MAP kinase and causes little or no enhanced agonist-stimulated arachidonate release from intact cells. Taken together, these data indicate that MAP kinase mediates, at least in part, the agonist-induced activation of cPLA2.