EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from the fungicide bupirimate.
PMID 8381343
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31510
Bupirimate, PESTANAL®, analytical standard
C13H24N4O3S