EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Effect of thioperamide, a histamine H3 receptor antagonist, on electrically induced convulsions in mice.
PMID 8386092
Abstract
The effect of thioperamide, a histamine H3 receptor antagonist, on electrically induced convulsions was studied in mice. Thioperamide significantly and dose dependently decreased the duration of each phase of convulsion and raised the electroconvulsive threshold. Its anticonvulsant effects were prevented by pretreatment with (R)-alpha-methylhistamine, a histamine H3 receptor agonist. These findings suggest that the effect of thioperamide on electrically induced convulsions is due to an increase in endogenous histamine release in the brain, an effect mediated by histamine H3 receptors. The anticonvulsant effect of thioperamide was antagonized strongly by mepyramine (or pyrilamine), a centrally acting histamine H1 receptor antagonist, but not by zolantidine, a centrally acting histamine H2 receptor antagonist. Thus, the blockade by mepyramine of the thioperamide-induced decrease in seizure susceptibility indicates that histamine released by thioperamide from the histaminergic nerve terminals interacts with the histamine H1 receptors of postsynaptic neurons. These findings support the hypothesis that the central histaminergic system is involved in the inhibition of seizures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T123
Thioperamide maleate salt, solid
C15H24N4S · C4H4O4