EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Metabolic labeling of mitogen-activated protein kinase kinase in A431 cells demonstrates phosphorylation on serine and threonine residues.
PMID 8389470
Abstract
Mitogen-activated protein (MAP) kinase kinase is an enzyme that activates the growth factor-regulated MAP kinase in vitro by a mechanism that involves direct phosphorylation of MAP kinase on tyrosine and threonine residues. MAP kinase kinase is stimulated by growth factor treatment of cells and has been shown to be inactivated with protein phosphatases, suggesting that it is regulated by protein phosphorylation. Analysis of two epidermal growth factor-stimulated forms of MAP kinase kinase, purified from 32P-labeled A431 cells, shows that the kinase is phosphorylated on serine and threonine residues and that treatment with protein phosphatases leads to serine dephosphorylation. Under conditions that lead to complete inactivation, only partial dephosphorylation of MAP kinase kinase is observed. Consistent with this finding, inactive forms of MAP kinase kinase, which separate from active forms during the course of purification, are also observed to be phosphorylated in intact cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
A431 Cell Line human, squamous carcinoma