EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Heat inactivation of paraoxonase and arylesterase activities in human and rabbit serum.
PMID 8393733
Abstract
The heat inactivation of esterases in human and rabbit serum was followed at 50 and 55 degrees C by measuring the decrease of activity with paraoxon, phenylacetate and beta-naphthylacetate as substrates. The rate of inactivation measured with the three substrates was slightly, but significantly different, indicating that the substrates are hydrolysed by different enzymes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70910
2-Naphthaleneacetic acid, purum, ≥98.0% (T)
C12H10O2
N4002
2-Naphthaleneacetic acid, 99%
C12H10O2