EMAIL THIS PAGE TO A FRIEND
Acta biochimica Polonica
Contribution of N-oxidation and .OH radicals to mutagenesis of 2-chloropyridine in Salmonella typhimurium.
PMID 8396828
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C69802
2-Chloropyridine, 99%
C5H4ClN