EMAIL THIS PAGE TO A FRIEND
International journal of legal medicine
Suicidal yew leave ingestion--phloroglucindimethylether (3,5-dimethoxyphenol) as a marker for poisoning from Taxus baccata.
PMID 8398891
Abstract
In a case of suicide in a depressive 19-year-old man with considerable ingestion of new leaves, resorption of yew ingredients could be demonstrated. The main substance could be identified as 3,5-dimethoxyphenol, the aglycone of taxicatine, which is a typical ingredient of yew leaves. 3,5-dimethoxyphenol was demonstrated in harvested yew leaves, stomach content and cardiac blood of the victim. Structure confirmation was achieved by means of HPLC, UV, GC-MS, IR and 1H-NMR spectroscopy. None of the Taxus alkoids could be identified. The components detected by TLC have not yet been identified. The results demonstrate that 3,5-dimethoxyphenol can be used as a marker in cases of intoxication by yew ingredients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
132632
3,5-Dimethoxyphenol, 99%
C8H10O3