EMAIL THIS PAGE TO A FRIEND
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis
Peritoneal membrane plasmapheresis.
PMID 8399678
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53310
Histamine bisphosphate monohydrate, ≥99.0% (NT)
C5H9N3 · 2H3PO4 · H2O
H7375
Histamine bisphosphate monohydrate
C5H9N3 · 2H3PO4 · H2O