EMAIL THIS PAGE TO A FRIEND
General and comparative endocrinology
Effects of natural, synthetic, aromatizable, and nonaromatizable androgens in inducing male sex differentiation in genotypic female chinook salmon (Oncorhynchus tshawytscha).
PMID 8405891
Abstract
The relative potency of several androgens to induce the male phenotype in sexually undifferentiated genotypic female chinook salmon were compared in two separate experiments. The aromatizable and nonaromatizable androgens testosterone (T) and 11-ketotestosterone (11-KT), and the synthetic aromatizable and nonaromatizable androgens 17 alpha-methyltestosterone (MT) and 17 alpha-methyldihydrotestosterone (MDHT) were administered to newly hatched alevins in a single 2-hr immersion treatment at doses ranging from 3.2 micrograms/liter to 10 mg/liter. The influence of these treatments on sex differentiation was evaluated by the histological examination of the resulting gonads 6 and 11 months later. In the control group, which was not exposed to exogenous steroids, no males or intersex fish were observed. In contrast, essentially 100% masculinization occurred in groups exposed to MDHT at dosages of 400 micrograms/liter and higher. Treatment with the aromatizable androgen MT resulted in a dose-dependent masculinization, with the production of 100% males at 400 micrograms/liter. However, higher doses resulted in fewer males. 11-KT and T were less potent than the synthetic androgens. The number of males produced after treatment with 11-KT followed a dose-dependent pattern while T showed virtually no masculinizing effect in inducing male phenotype in these studies. The resultant AD50 dosage levels (dosage at which 50% of the genotypic females were sex-reversed into phenotypic males) after a single 2-hr immersion treatment were: 30, 60, and 500 micrograms/liter for MDHT, MT, and 11-KT, respectively.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5626
17α-Methylandrostan-17β-ol-3-one, analytical standard
C20H32O2