EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Cytogenetic activity of 2,4-D dimethylamine salt and ammonium nitrate in isolated and combined exposure of plants in experiment].
PMID 8406091
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10612 2,4-D dimethylamine salt, analytical standard