EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Cloning of a human acid sphingomyelinase cDNA with a new mutation that renders the enzyme inactive.
PMID 8407868
Abstract
A cDNA encoding human acid sphingomyelinase was initially obtained by screening a placental cDNA library in lambda gt11 with a synthetic oligonucleotide probe and subsequently with partial cDNA. The full-length cDNA, hPSM55, comprised 2,376 nucleotides, with a 5' untranslated sequence of 122 nucleotides, an open reading frame of 1,884 nucleotides encoding a protein of 627 amino acids, and a 3' untranslated region of 370 bases. hPSM55 was almost identical to pASM-1FL reported by Schuchman et al. (J. Biol. Chem. 266, 8531-8539, 1991) except for a 6 base pair deletion in the signal peptide, which indicated the possible removal of valine and leucine residues between positions 36 and 37, and a 463T- to C-transition, which indicated a possible substitution of 155arginine for cystine. This cDNA was expressed in both COS-7 cells and Chinese hamster ovary cells. There was no increase in acid sphingomyelinase activity in either cell line following transfection. However, the correction of a single base change, 463C to T, in hPSM55 caused increased acid sphingomyelinase activity in transfectants. These results suggest that the mutation of nucleotide 463C to T plays an important role in the catalytic activity of acid sphingomyelinase.