EMAIL THIS PAGE TO A FRIEND
The Journal of pediatrics
Molybdenum cofactor deficiency.
PMID 8410516
Abstract
We describe a new case of molybdenum cofactor deficiency, an underrecognized inborn error of metabolism that results in neonatal seizures and neurologic abnormalities. Characteristic biochemical defects in affected individuals include hypouricemia, elevated urine sulfate (detectable by dipstick), and elevated S-sulfocysteine (detectable by anion exchange chromatography). This disorder should be considered in the differential diagnosis of neonatal seizures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2196
L-Cysteine S-sulfate, ≥98% (TLC)
C3H7NO5S2