EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Characterization of a new human diploid cell strain, IMR-90.
PMID 841339
Abstract
A new human diploid fibroblast-like cell line has been established from lung tissue of a female fetus. This has been frozen away in large quantity and characterized for use in research and related purposes. This is the first of a planned series of human cell lines to be established, characterized, and banked in large quantity in support of the National Institute on Aging research and general cell biology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
IMR-90 Cell Line human, 85020204