EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[Absorption and excretion of apigenin, apigenin-7-glycoside and herniarin after oral administration of extracts of Matricaria recutita (L.) (syn. Chamomilla recutita (L.) Rauschert)].
PMID 8415854