EMAIL THIS PAGE TO A FRIEND
Glia
Characterization of microglia and macrophages in the central nervous system of rats: definition of the differential expression of molecules using standard and novel monoclonal antibodies in normal CNS and in four models of parenchymal reaction.
PMID 8423060