EMAIL THIS PAGE TO A FRIEND
Dermatology (Basel, Switzerland)
Diflorasone diacetate ointment 0.05% versus betamethasone dipropionate ointment 0.05% in moderate-severe plaque-type psoriasis.
PMID 8428041
Abstract
We report the results of a 2-week double-blind, parallel clinical trial comparing two superpotent topical glucocorticosteroid ointments, diflorasone diacetate 0.05% and betamethasone dipropionate 0.05%, in psoriatic adults. Both corticosteroid ointments were fast acting and highly efficacious. 40 of the 44 patients who initially enrolled completed the trial. There were no statistically significant differences between the two glucocorticoids with respect to erythema, scaling, induration or the investigator's global evaluation after either 1 or 2 weeks of therapy. The level of patient satisfaction with the efficacy and cosmetic acceptability of these two category I glucocorticoids was similar. No systemic or local adverse reactions were noted.