EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Efficient DNA transfection of quiescent mammalian cells using poly-L-ornithine.
PMID 8441700
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3655 Poly-L-ornithine hydrobromide, mol wt 30,000-70,000