EMAIL THIS PAGE TO A FRIEND
Nihon Naibunpi Gakkai zasshi
[Analysis of corticoids in adrenal glands by high performance liquid chromatography (HPLC)].
PMID 8449245
Abstract
The HPLC system was used to separate and measure 10 kinds of corticoids in adrenal tissues. Calibration curves were drawn as straight lines that ranged from 1.25 to 20ng, or 1.25 to 200ng by peak area calculated with the chromatointegrator. The samples for the assay were extracted from homogenized tissues and treated with methanol to remove non-steroidal contaminants which may interfere with the ultraviolet absorption monitor. The recovery rate during the assay procedure was calculated using testosterone as the internal standard, because testosterone was not detected in any adrenal tissue examined in the present study. Contents of corticoids were measured in normal adrenal glands obtained during radical nephrectomy for renal cancer and in functioning adrenal adenomas. Steroid levels in the adrenal glands and tumors have been measured by radioimmunoassay until now, and the data obtained in the present study were compared with those in previous reports. Main steroids in normal adrenals were cortisol (F) and corticosterone (B), and there were certain amounts of 11-deoxycortisol (S), 11-deoxycorticosterone (DOC) and precursor steroids. 11 beta-hydroxy-androstenedione was the main androgen in the adrenal gland. Mineralocorticoids other than B and DOC were very low in the normal adrenals. There was a certain balance between the production of cortisol and corticosterone in normal adrenals. In functioning adenomas, the levels of F, B and aldosterone, and F to B ratios (F/B) varied according to their biological features. Although with the HPLC system it was possible to obtain the production balance of each steroid clearly in the chromatogram, we could not detect the delta 5-3 hydroxysteroids such as pregnenolone and dehydroepiandrosterone using the ultraviolet absorption monitor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3009
4-Androsten-11β-ol-3,17-dione
C19H26O3