EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact allergy to benzyl alcohol and benzyl paraben.
PMID 8462301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
380709
Benzyl 4-hydroxybenzoate, 99%
C14H12O3