EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Syntheses of the potent inhibitors of neuraminidase, N-(1,2-dihydroxypropyl) derivatives of siastatin B and its 4-deoxy analogs.
PMID 8468246
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8063
Siastatin B, lyophilized powder, from microbial
C8H14N2O5