EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Direct high-performance liquid chromatographic determination in urine of the enantiomers of propranolol and its major basic metabolite 4-hydroxypropranolol.
PMID 8468379
Abstract
A method is described for quantitation of underivatized enantiomers of propranolol and its major basic metabolite, 4-hydroxypropranolol, in urine samples by high-performance liquid chromatography with fluorescence detection, using a cellulose tris(3,5-dimethylphenylcarbamate) chiral stationary phase. This method was found to be precise and accurate for the measurement of propranolol and 4-hydroxypropranolol concentrations in urine from pharmacokinetic investigations. This method represents the first assay for direct determination of 4-hydroxypropranolol enantiomers. The ability to easily measure 4-hydroxypropranolol enantiomers is valuable because the stereoselective disposition of propranolol is primarily due to stereoselective metabolism in the pathway responsible for generation of 4-hydroxypropranolol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00881
(±)-4-Hydroxypropranolol hydrochloride, analytical standard
C16H21NO3·HCl