EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A highly sensitive and fast nonradioactive method for detection of polymerase chain reaction products.
PMID 8470800
Abstract
A novel system for the detection of polymerase chain reaction (PCR) products has been developed. The system is based on a PCR in which one of the primers is biotinylated and digoxigenin-11-dUTP is incorporated during elongation. Biotinylated PCR products are captured on streptavidin-coated solid supports, and alkaline phosphatase conjugated to anti-digoxigenin antibody is subsequently bound to the incorporated digoxigenin. The detection may be obtained with colorimetric, fluorescent, or luminescent substrates for alkaline phosphatase. The detection system can be performed in microtiter plates allowing easy handling of multiple samples. The total assay time following the PCR is between 1 and 2 h dependent on the type of substrate and the type of solid support applied in the system. Within this period of time the system is capable of detecting 1 template in 29 cycles of PCR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8883
4-Methylumbelliferyl phosphate, phosphatase substrate
C10H9O6P