EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Determination of bromofos, diazinon and methylpyrimifos in fruit and vegetables].
PMID 847332
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33400
Bromophos-methyl, PESTANAL®, analytical standard
C8H8BrCl2O3PS