EMAIL THIS PAGE TO A FRIEND
FEBS letters
Inactivation of alpha-ketoglutarate dehydrogenase during oxidative decarboxylation of alpha-ketoadipic acid.
PMID 8495733
Abstract
alpha-Ketoglutarate dehydrogenase was inactivated irreversibly and completely during oxidation of alpha-ketoadipic acid. The inactivation was revealed both in the model system with ferricyanide and in the overall reaction catalyzed by the alpha-ketoglutarate dehydrogenase complex. Neither substrate depletion nor product accumulation induced the inactivation. The results obtained were compared with recent data on the enzyme inactivation during oxidation of alpha-ketoglutaric acid. The differences in the inactivation kinetics observed with the two substrates of the enzyme were analyzed. They seem not to reflect the different mechanisms of the inactivation, but, rather, depend on the changes in the rates of the individual stages of the process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75447
2-Oxoadipic acid, ≥95.0% (HPLC)
C6H8O5