EMAIL THIS PAGE TO A FRIEND
The Veterinary clinics of North America. Small animal practice
Hepatotoxicity associated with pharmacologic agents in dogs and cats.
PMID 8503165
Abstract
Historical aspects, mechanisms, and treatment of hepatotoxicity associated with pharmacologic agents in dogs and cats are discussed. All agents that cause clinically relevant drug-induced hepatotoxicity have been reviewed within the last 5 years; therefore, only selected drugs that more recently have been observed to cause hepatic injury in companion animals are described in detail. These include diethylcarbamazine/oxibendazole, phenobarbital, and trimethoprim/sulfadiazine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32924
Oxibendazole, VETRANAL®, analytical standard
C12H15N3O3
O3132
Oxibendazole, ~98%
C12H15N3O3