EMAIL THIS PAGE TO A FRIEND
Blood
Elevation of serum cystathionine levels in patients with cobalamin and folate deficiency.
PMID 8507876
Abstract
Homocysteine can be methylated to form methionine by the cobalamin- (Cbl) and folate-dependent enzyme, methionine synthase; serum levels of total homocysteine are elevated in greater than 95% of patients with either Cbl or folate deficiency. Homocysteine can also condense with serine to form cystathionine in a pyridoxal phosphate-dependent reaction catalyzed by cystathionine beta-synthase. Cystathionine is subsequently cleaved to cysteine and alpha-ketobutyrate by the pyridoxal phosphate-dependent enzyme gamma-cystathionase. To assess levels of cystathionine in Cbl and folate deficiency, we developed a new capillary gas chromatographic-mass spectrometric assay and measured cystathionine in the serum of normal subjects and patients with clinically confirmed deficiencies of these vitamins. The normal range for serum cystathionine was 65 to 301 nmol/L (median = 126 nmol/L) for 50 normal blood donors. In 30 patients with clinically confirmed Cbl deficiency, values for cystathionine ranged from 208 nmol/L to 2,920 nmol/L (median = 816 nmol/L) and 26 (87%) had levels above the normal range. In 20 patients with clinically confirmed folate deficiency, cystathionine concentrations ranged from 138 nmol/L to 4,150 nmol/L (median = 1,560 nmol/L) and 19 (95%) had values above the normal range. Five homozygotes for cystathionine beta-synthase deficiency had high values for serum-total homocysteine and low or low-normal values for serum cystathionine that ranged from 30 nmol/L to 114 nmol/L even though they were on treatment with pyridoxine and had partially responded. One patient with a defect in the synthesis of 5-CH3-tetrahydrofolate and five patients with defects in the synthesis of CH3-Cbl had high values for serum-total homocysteine and high values for cystathionine that ranged from 311 nmol/L to 1,500 nmol/L even though they were on treatment with folic acid and Cbl, respectively, and had partially responded. We conclude that levels of cystathionine are evaluated in the serum of most patients with Cbl and folate deficiency and that they are useful in the differential diagnosis of an elevated serum-total homocysteine level.