EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Preliminary observations on the in vitro toxicity of N-butylbenzenesulfonamide: a newly discovered neurotoxin.
PMID 8512189
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B90653
N-Butylbenzenesulfonamide, 99%
C10H15NO2S